Learn More
Bon Opus Biosciences Biotinylated Human FcRn Heterodimer (C-6His-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY30100UG
Encompass_Preferred
Biotinylated Recombinant Human IgG Fc fragment receptor transporter is produced by our Mammalian expression system and the target gene encoding Ala24-Ser297/Ile21-Met119 is expressed with a 6His, Avitag at the C-terminus. Expression system: Human Cells. Accession# AAF72596/P61769. Formulation: Lyophilized from a 0.2 μm filtered solution of 50mM HEPES, 150mM NaCl, pH 7.4. Bon Opus Cat# CY30
Catalog No. 50-208-0525