Learn More
Bon Opus Biosciences Biotinylated Mouse P-selectin (C-Avi-6His) -100ug
SDP
Supplier:  Bon Opus Biosciences CU68100UG
Encompass_Preferred
Biotinylated Recombinant Mouse P-selectin is produced by our Mammalian expression system and the target gene encoding Trp42­Ala709 is expressed with a Avi, 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q01102. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CU68
Catalog No. 50-208-0405