Learn More
ATCC BT142 mut/-; Glioma cell line; Human (Homo sapiens)
Supplier:  ATCC ACS1018
Encompass
BT142 mut/- is a glioma brain tumor stem cell line derived from the left frontal lobe of a 38-year-old, White male. BT142 mut/- contains a homozygous IDH1 R132H mutation, which originated from a heterozygous IDH1 R132H BT142 cells. The cells grow as phase-bright, smooth spheres. These neural cells can be used for your cancer, neuroscience, stem cell, and toxicology research.
Catalog No. 50-238-2028