Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BT2 10mM 1mL | 34576-94-8 | 247.10 g/mol | C9H4Cl2O2S | 1 ML
SDP

Catalog No. 5000229672
Encompass_Preferred

BT2 (Benzo[b]thiophene-2-carboxylic acid, 3,6-dichloro-) is a small-molecule inhibitor of branched-chain alpha-ketoacid dehydrogenase kinase (BCKDC kinase/BDK) supplied for research use. It is available as a 10 mM solution in DMSO and as a solid, and is intended for biochemical and cellular studies (not for human use).

  • Chemical name: Benzo[b]thiophene-2-carboxylic acid, 3,6-dichloro-.
  • Available as 10 mM solution in DMSO (1 mL) and as solid in multiple sizes.
  • High reported purity (around 99%).
  • Molecular weight: 247.10 g/mol; formula: C9H4Cl2O2S.
  • Recommended storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-29672 Supplier Medchemexpress LLC Supplier No. HY11485510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BT2 (Benzo[b]thiophene-2-carboxylic acid, 3,6-dichloro-) is a small-molecule inhibitor of branched-chain alpha-ketoacid dehydrogenase kinase (BCKDC kinase/BDK) supplied for research use. It is available as a 10 mM solution in DMSO and as a solid, and is intended for biochemical and cellular studies (not for human use).

  • Chemical name: Benzo[b]thiophene-2-carboxylic acid, 3,6-dichloro-.
  • Available as 10 mM solution in DMSO (1 mL) and as solid in multiple sizes.
  • High reported purity (around 99%).
  • Molecular weight: 247.10 g/mol; formula: C9H4Cl2O2S.
  • Recommended storage: powder -20°C (up to 3 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.