Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BT2 5mg | 34576-94-8 | 247.10 g/mol | C9H4Cl2O2S | 5 MG
SDP

Catalog No. 5000229673
Encompass_Preferred

BT2 (3,6-dichloro-1-benzothiophene-2-carboxylic acid; CAS 34576-94-8) is a small-molecule inhibitor of the branched-chain α-ketoacid dehydrogenase complex kinase (BCKDC/BDK), supplied as a research-grade powder for biochemical and cell-based studies.

  • Off-white to yellow powder, available as 5 MG pack
  • Chemical formula C9H4Cl2O2S; molecular weight 247.10 g/mol
  • Reported purity varies by supplier (examples range from ~99.8% to ≥95%)
  • Storage: powder at -20°C for long-term; in solvent, -80°C up to 6 months or -20°C up to 1 month
  • Soluble in DMSO; for research use only

Catalog No. 50-002-29673 Supplier Medchemexpress LLC Supplier No. HY1148555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BT2 (3,6-dichloro-1-benzothiophene-2-carboxylic acid; CAS 34576-94-8) is a small-molecule inhibitor of the branched-chain α-ketoacid dehydrogenase complex kinase (BCKDC/BDK), supplied as a research-grade powder for biochemical and cell-based studies.

  • Off-white to yellow powder, available as 5 MG pack
  • Chemical formula C9H4Cl2O2S; molecular weight 247.10 g/mol
  • Reported purity varies by supplier (examples range from ~99.8% to ≥95%)
  • Storage: powder at -20°C for long-term; in solvent, -80°C up to 6 months or -20°C up to 1 month
  • Soluble in DMSO; for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.