Learn More
Alkali Scientific Alkali Scientific™ BT3001 Orbi-Shaker Orbital Shaker with 13 x 12 inch platform 115V
SDP
Catalog No. 503029560
Encompass_Preferred
Alkali Scientific™ BT3001 Orbi-Shaker Orbital Shaker with non-slip, rubber mat platform. • High capacity, up to 9 x 500ml, 5 x 1L or 2 x 2L• Horizontally circular 19mm orbit for aeration & mixing• Digital speed control up to 300 rpm• Instantly exchange flask clamps with MAGic Clamp™ accessoriesThe powerful Orbi-Shaker is capable of mixing a 14lb load capacity up to speeds as high as 300 rpm. The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays. The convenient MAGic Clamp platform (sold separately) is also available for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.
Catalog No. 50-302-9560 Supplier Alkali Scientific Supplier No. RS7061
Alkali Scientific™ BT3001 Orbi-Shaker Orbital Shaker with non-slip, rubber mat platform. • High capacity, up to 9 x 500ml, 5 x 1L or 2 x 2L• Horizontally circular 19mm orbit for aeration & mixing• Digital speed control up to 300 rpm• Instantly exchange flask clamps with MAGic Clamp™ accessoriesThe powerful Orbi-Shaker is capable of mixing a 14lb load capacity up to speeds as high as 300 rpm. The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays. The convenient MAGic Clamp platform (sold separately) is also available for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.