Learn More
Alkali Scientific BT4001 Orbi-Shaker CO2 with To
SDP
Catalog No. 503029638
Encompass_Preferred
The new Alkali ScientificBT4001 Orbi-Shaker is designed for shaking mixing aeration applications in extreme environments including CO2 Shaking Incubator It can withstand temperatures from 0 to 60C and humidity levels up to 100% To avoid contamination unintentional release of gas or deviations from protocol all settings are adjusted via the included remote control module This module can be stored outside of the chamber and connects to the shaker via a flat (1mm thickness) cable that easily passes through a port or underneath an incubator door without compromising the seal This allows for user adjustment of run time or mixing speed without opening the chamber door(s) The instrument is supplied with a non-slip rubber mat platform In addition convenient MAGic Clampplatforms (of multiple sizes) are also available for use with Erlenmeyer flasks and test tube racks The unique magnetic attachment method (PATENTED) has been widely recognized as the easiest way to instantly change between
Catalog No. 50-302-9638 Supplier Alkali Scientific Supplier No. RS7505
The new Alkali ScientificBT4001 Orbi-Shaker is designed for shaking mixing aeration applications in extreme environments including CO2 Shaking Incubator It can withstand temperatures from 0 to 60C and humidity levels up to 100% To avoid contamination unintentional release of gas or deviations from protocol all settings are adjusted via the included remote control module This module can be stored outside of the chamber and connects to the shaker via a flat (1mm thickness) cable that easily passes through a port or underneath an incubator door without compromising the seal This allows for user adjustment of run time or mixing speed without opening the chamber door(s) The instrument is supplied with a non-slip rubber mat platform In addition convenient MAGic Clampplatforms (of multiple sizes) are also available for use with Erlenmeyer flasks and test tube racks The unique magnetic attachment method (PATENTED) has been widely recognized as the easiest way to instantly change between