Learn More
Alkali Scientific Alkali Scientific™ (BT4011) Orbi-Shaker CO2 Extra Large Platform with Remote Controller for CO2 Incubators, 115V
SDP
Catalog No. 503029538
Encompass_Preferred
Designed to fit and maximize capacity in most CO2 incubators Remote control, adjust without disrupting the CO2 environment Horizontally circular 19mm orbit for aeration & mixing Instantly exchange flask clamps with MAGic Clamp™ accessories (PATENTED) The newly upgraded Alkali Scientific™ Orbi-Shaker CO2 XL (BT4011) now features an advanced touch screen controller for more powerful programming. The BT4011 Orbi-Shaker CO2 XL is supplied with a 16 x 16.5" non-slip platform.If you will be shaking at speeds below 120 rpm you can use the included rubber topped platform. For speeds in excess of this the convenient MAGic Clamp™ platform is available for use with Erlenmeyer flasks and test tube racks.These magnetic platform and clamps make is easy to swap out one size for another. 
Catalog No. 50-302-9538 Supplier Alkali Scientific Supplier No. RS7065
Designed to fit and maximize capacity in most CO2 incubators Remote control, adjust without disrupting the CO2 environment Horizontally circular 19mm orbit for aeration & mixing Instantly exchange flask clamps with MAGic Clamp™ accessories (PATENTED) The newly upgraded Alkali Scientific™ Orbi-Shaker CO2 XL (BT4011) now features an advanced touch screen controller for more powerful programming. The BT4011 Orbi-Shaker CO2 XL is supplied with a 16 x 16.5" non-slip platform.If you will be shaking at speeds below 120 rpm you can use the included rubber topped platform. For speeds in excess of this the convenient MAGic Clamp™ platform is available for use with Erlenmeyer flasks and test tube racks.These magnetic platform and clamps make is easy to swap out one size for another. 