Learn More
Rarecyte RareCyte RarePlex BT-474 spike-in CTC Control Slide, 10-Pack
Catalog No. NC1854147
Encompass
A pack of ten, batch control slides to be stained, scanned, and reviewed with experimental slides. Control slides contain a monolayer of buffy coat and spiked-in BT-474 cells (epithelial cells from human breast carcinoma). This control slide is used to assess the performance of the immunofluorescent staining process for the detection of CTCs. Cells may be identified by biomarkers: Nuclear DNA, CK/EpCAM, CD45, HER2 and ER. Compatible kits: RareCyte RarePlex HER2/ER CTC Panel Kit, Ventana (SYTOX) (24-1257-002); CTC Panel Kit, Ventana (SYTOX) (24-1245-002); CTC Panel Kit, Leica (SYTOX) (24-1245-001); CTC Panel Kit, Ventana (Hoechst) (24-1277-002); CTC Panel Kit, Leica (Hoechst) (24-1277-001); CTC Panel Kit, Manual Staining (Hoechst) (24-1277-004); PD-L1 CTC Panel Kit, Ventana (24-1280-002); Developer Kit: 488 nm, Leica (28-1001-481); 405 nm, Leica (28-1001-401); 405 nm, Ventana (28-1001-402); 488 nm, Ventana(28-1001-482) and Cell Discovery Kit, Leica (24-1294-001).
Catalog No. NC1854147 Supplier Rarecyte Supplier No. 241247000
A pack of ten, batch control slides to be stained, scanned, and reviewed with experimental slides. Control slides contain a monolayer of buffy coat and spiked-in BT-474 cells (epithelial cells from human breast carcinoma). This control slide is used to assess the performance of the immunofluorescent staining process for the detection of CTCs. Cells may be identified by biomarkers: Nuclear DNA, CK/EpCAM, CD45, HER2 and ER. Compatible kits: RareCyte RarePlex HER2/ER CTC Panel Kit, Ventana (SYTOX) (24-1257-002); CTC Panel Kit, Ventana (SYTOX) (24-1245-002); CTC Panel Kit, Leica (SYTOX) (24-1245-001); CTC Panel Kit, Ventana (Hoechst) (24-1277-002); CTC Panel Kit, Leica (Hoechst) (24-1277-001); CTC Panel Kit, Manual Staining (Hoechst) (24-1277-004); PD-L1 CTC Panel Kit, Ventana (24-1280-002); Developer Kit: 488 nm, Leica (28-1001-481); 405 nm, Leica (28-1001-401); 405 nm, Ventana (28-1001-402); 488 nm, Ventana(28-1001-482) and Cell Discovery Kit, Leica (24-1294-001).