Learn More
ATCC BT-549 Quantitative; purified DNA, 3 ug; Human
Catalog No. 502382984
Encompass
Quantified for oncology biomarker TP53 p.R249S via NGS and ddPCR
Catalog No. 50-238-2984 Supplier ATCC Supplier No. HTB122DQ
Quantified for oncology biomarker TP53 p.R249S via NGS and ddPCR