Learn More
New England Biolabs, Inc. BtsI-v2 – 500 units
SDP
Catalog No. NC0830981
Encompass
BtsI-v2 is an engineered and improved version of BtsI that recognizes the sequence GCAGTGNN^.
Catalog No. NC0830981 Supplier New England Biolabs, Inc. Supplier No. R0667S
BtsI-v2 is an engineered and improved version of BtsI that recognizes the sequence GCAGTGNN^.