Learn More
Medchemexpress LLC HY-15877 5mg Medchemexpress, BTB06584 CAS:219793-45-0 Purity:>98%
SDP
Catalog No. 502081814
Encompass_Preferred
Medchemexpress, HY-15877 5mg BTB06584 CAS:219793-45-0 BTB06584 is a selective and IF1-dependent mitochondrial F1Fo-ATPase inhibitor without compromising ATP synthesis. BTB06584 can delays ischaemic cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1814 Supplier Medchemexpress LLC Supplier No. HY158775MG
Medchemexpress, HY-15877 5mg BTB06584 CAS:219793-45-0 BTB06584 is a selective and IF1-dependent mitochondrial F1Fo-ATPase inhibitor without compromising ATP synthesis. BTB06584 can delays ischaemic cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.