Learn More
Strem, An Ascensus Company CAS# [186598-40-3]. 5g. Bis(t-butylamino)silane, BTBAS (99.999%-Si) PURATREM.
SDP
Supplier:  Strem, An Ascensus Company 1410725G
Encompass
CAS# 186598-40-3. 5g. Bis(t-butylamino)silane, BTBAS (99.999%-Si) PURATREM. . Molecular Weight: 174.36. Molecular Formula: C8H22N2Si. Color/form: colorless liq. Strem# 14-1072. www.strem.com/catalog/v3/14-1072/
Catalog No. 50-197-9361