Learn More
Aviva Systems Biology Corp BTBD10 Antibody - N-terminal region (ARP35376_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35376_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTBD10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BTBD10. Alternate names: GMRP-1, GMRP1, MGC13007ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55718