Learn More
Aviva Systems Biology Corp BTBD14B Antibody - middle region (ARP34614_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34614_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BTBD14B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human BTBD14B. Alternate names: NAC1, BEND8, NAC-1, BTBD14BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-52968
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.