Learn More
Aviva Systems Biology Corp BTBD14B Antibody - middle region (ARP34653_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34653_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTBD14B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BTBD14B. Alternate names: FLJ37383, NAC1, BEND8, NAC-1, BTBD14BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55502