Learn More
Aviva Systems Biology Corp BTBD2 Antibody - N-terminal region (ARP34458_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34458_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTBD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BTBD2. Alternate names: -ApplicationWB. Species reactivity: Cow, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55400