Learn More
Aviva Systems Biology Corp BTBD3 Antibody - middle region (ARP34740_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34740_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BTBD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BTBD3. Alternate names: dJ742J24.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55540