Learn More
Aviva Systems Biology Corp BTBD3 Antibody - N-terminal region (ARP34341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTBD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BTBD3. Alternate names: KIAA0952, MGC130038, MGC130039, dJ742J24.1ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-55349