Learn More
Aviva Systems Biology Corp BTBD5 Antibody - middle region (ARP34570_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34570_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BTBD5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BTBD5. Alternate names: BTBD5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55455