Learn More
Novus Biologicals™ BTBD7 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 55727 |
| Species | Human |
| Purity | >85%, by HPLC |
| Concentration | 2.5 mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for Antibody NBP1-49652 Applications: Immmunodepletion, Competition Assay |
| Gene Alias | BTB (POZ) domain containing 7, DKFZp686N0544, FLJ10648, FUP1BTB/POZ domain-containing protein 7, KIAA1525, MGC48310 |
| Gene Symbol | BTBD7 |
| Quantity | 0.25 mg |
| Research Category | Cancer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.