Learn More
Bellco Glass, Inc. BTBII Recprocating Tray,10x10"
SDP
Catalog No. 501945993
Encompass_Preferred
This tray only fits the back-to-basics II reciprocating shaker units. Includes rubber mat with peg and grommet design. Customer must use this tray or stainless steel tray 7677-32010 for reciprocating shaker units. MADE IN THE USA Size: 10"x10"
Catalog No. 50-194-5993 Supplier Bellco Glass, Inc. Supplier No. 767731010
This tray only fits the back-to-basics II reciprocating shaker units. Includes rubber mat with peg and grommet design. Customer must use this tray or stainless steel tray 7677-32010 for reciprocating shaker units. MADE IN THE USA Size: 10"x10"