Learn More
Strem, An Ascensus Company CAS# [406462-43-9]. 1g. Bis(t-butylimido)bis(dimethylamino)tungsten(VI), min. 97% BTBMW. MFCD07369712
SDP
Supplier:  Strem, An Ascensus Company 7412251G
Encompass
CAS# [406462-43-9]. 1g. Bis(t-butylimido)bis(dimethylamino)tungsten(VI), min. 97% BTBMW. MFCD07369712. Molecular Weight: 414.23. Molecular Formula: C12H30N4W. Color/form: yellow to orange liq. Strem# 74-1225. www.strem.com/catalog/v3/74-1225/
Catalog No. 50-197-9381