Learn More
Protexer Corporation Shoe Cover Refill - Non-skid, Non-woven, Polypropylene - 40g - Case - 600 pairs
Catalog No. NC0950035
Encompass_Preferred
Constructed of durable spunbond polypropylene material (Density: 40 grams / m2) with non-skid, non-conductive soles. Skid-resistant pattern improves friction and traction. Strong elastic band around ankle allows for a universal fit. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.
This item is not returnable. View return policy
Catalog No. NC0950035 Supplier Protexer Corporation Supplier No. KBNS40600
Constructed of durable spunbond polypropylene material (Density: 40 grams / m2) with non-skid, non-conductive soles. Skid-resistant pattern improves friction and traction. Strong elastic band around ankle allows for a universal fit. One size fits most (up to men’s size 15). Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.