Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ BTD Recombinant Protein

Catalog No. 89009930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-009-930 10 μg
89-009-931 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-009-930 Supplier Abnova™ Supplier No. H00000686P0110
Add to Cart
Add to Cart

Human BTD full-length ORF recombinant protein with GST-tag at N-terminal

Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency.

  • Theoretical MW (kDa): 85.47
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MAHAHIQGGRRAKSRFVVCIMSGARSKLALFLCGCYVVALGAHTGEESVADHHEAEYYVAAVYEHPSILSLNPLALISRQEALELMNQNLDIYEQQVMTAAQKDVQIIVFPEDGIHGFNFTRTSIYPFLDFMPSPQVVRWNPCLEPHRFNDTEVLQRLSCMAIRGDMFLVANLGTKEPCHSSDPRCPK
DGRYQFNTNVVFSNNGTLVDRYRKHNLYFEAAFDVPLKVDLITFDTPFAGRFGIFTCFDILFFDPAIRVLRDYKVKHVVYPTAWMNQLPLLAAIEIQKAFAVAFGINVLAANVHHPVLGMTGSGIHTPLESFWYHDMENPKSHLIIAQVAKNPVGLIGAENATGETDPSHSKFLKILSGDPYCEKDAQEVHCDEATKWNVNAPPTFHSEMMYDNFTLVPVWGKEGYLHVCSNGLCCYLLYERPTLSKELYALGVFDGLHTVHGTYYIQVCALVRCGGLGFDTCGQEITEATGIFEFHLWGNFSTSYIFPLFLTSGMTLEVPDQLGWENDHYFLRKSRLSSGLVTAALYGRLYERD

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH12099
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 686
Molecular Weight (g/mol) 85.47
Name BTD (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Common Name BTD
Gene Symbol BTD
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.