Learn More
Medchemexpress LLC HY-118266 5mg Medchemexpress, BTdCPU CAS:1257423-87-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1182665MG
Encompass_Preferred
Medchemexpress, HY-118266 5mg BTdCPU CAS:1257423-87-2 BTdCPU is a potent heme-regulated eIF2α kinase (HRI) activator. BTdCPU promotes eIF2α phosphorylation and induced apoptosis in resistant cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9501