Learn More
Quality Biological Inc Bottle Top Filter, 150ml PES, 0.22um, 24/CS
SDP
Supplier:  Quality Biological Inc P886229716
Encompass_Preferred
Bottle Top Filter, 150ml PES, 0.22um, 24/CS
Catalog No. 50-751-7791