Learn More
Aviva Systems Biology Corp BTG2 Antibody - N-terminal region (ARP33560_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33560_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BTG2. Alternate names: MGC126063, MGC126064, PC3, TIS21ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55071