Learn More
Aviva Systems Biology Corp BTG2 Antibody - middle region (ARP33561_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33561_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human BTG2. Alternate names: MGC126063, MGC126064, PC3, TIS21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-55072