Learn More
Aviva Systems Biology Corp BTG4 Antibody - middle region (ARP56981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BTG4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BTG4. Alternate names: MGC33003, PC3BApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62053