Learn More
New England Biolabs, Inc. BtgI – 1000 units
SDP
Catalog No. 50812338
Encompass
A restriction endonuclease that recognizes the sequence C^CRYG_G.
Catalog No. 50-812-338 Supplier New England Biolabs, Inc. Supplier No. R0608S
A restriction endonuclease that recognizes the sequence C^CRYG_G.