Learn More
New England Biolabs, Inc. BtgZI – 500 units
SDP
Catalog No. 50812411
Encompass
A restriction endonuclease that recognizes the sequence GCGATGNNNNNNNNNN^NNNN_.
Catalog No. 50-812-411 Supplier New England Biolabs, Inc. Supplier No. R0703L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence GCGATGNNNNNNNNNN^NNNN_.