Learn More
New England Biolabs, Inc. BtgZI – 100 units
SDP
Catalog No. 50812412
Encompass
A restriction endonuclease that recognizes the sequence GCGATGNNNNNNNNNN^NNNN_.
Catalog No. 50-812-412 Supplier New England Biolabs, Inc. Supplier No. R0703S
A restriction endonuclease that recognizes the sequence GCGATGNNNNNNNNNN^NNNN_.