Learn More
Sino Biological Recombinant Human RNF130 & BRAF Protein (GST Tag), Active
Catalog No. 502575787
Encompass_Preferred
Recombinant human fusion RNF130 (1-231 exon3)-BRAF (381-end or exon9-18) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5787 Supplier Sino Biological Supplier No. B0819GG50
Recombinant human fusion RNF130 (1-231 exon3)-BRAF (381-end or exon9-18) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.