Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bruton's tyrosine kinase (BTK), human, C481S, biotinylated | >90.0% | 50UG
SDP

Catalog No. 5000175950
Encompass_Preferred

Recombinant human Bruton's tyrosine kinase (BTK) C481S mutant supplied as an active, site-specifically biotinylated protein with N-terminal His and Avi tags. Expressed in Sf9 insect cells and provided as a 50 μg formulation in a filtered buffered solution. Purity is greater than 90% and molecular weight is approximately 80.4 kDa; store at -80°C and avoid repeated freeze-thaw cycles.

  • Expressed in Sf9 insect cells.
  • Site-specific biotinylation with N-terminal His and Avi tags.
  • Active enzyme activity confirmed by off-chip mobility shift assay.
  • Contains C481S mutation to model inhibitor resistance.
  • Supplied as a 50 μg aliquot in filtered buffered solution.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Store at -80°C; stable short-term at -20°C after opening; avoid repeated freeze-thaw.

Catalog No. 50-001-75950 Supplier Medchemexpress LLC Supplier No. HYP70273050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Bruton's tyrosine kinase (BTK) C481S mutant supplied as an active, site-specifically biotinylated protein with N-terminal His and Avi tags. Expressed in Sf9 insect cells and provided as a 50 μg formulation in a filtered buffered solution. Purity is greater than 90% and molecular weight is approximately 80.4 kDa; store at -80°C and avoid repeated freeze-thaw cycles.

  • Expressed in Sf9 insect cells.
  • Site-specific biotinylation with N-terminal His and Avi tags.
  • Active enzyme activity confirmed by off-chip mobility shift assay.
  • Contains C481S mutation to model inhibitor resistance.
  • Supplied as a 50 μg aliquot in filtered buffered solution.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Store at -80°C; stable short-term at -20°C after opening; avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.