Learn More
Selleck Chemical LLC BTK inhibitor 1
SDP
Catalog No. 5000000819
Encompass_Preferred
BTK inhibitor 1 (compound 27) is an inhibitor of BTK with an IC50 of 0 11 nM for Btk and inhibits B cell activation in hWB with an IC50 of 2 nM
Catalog No. 50-000-00819 Supplier Selleck Chemical LLC Supplier No. S86792MG
BTK inhibitor 1 (compound 27) is an inhibitor of BTK with an IC50 of 0 11 nM for Btk and inhibits B cell activation in hWB with an IC50 of 2 nM