Learn More
TARGETMOL CHEMICALS INC BTK INHIBITOR 1 5MG
SDP
Catalog No. 502580224
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BTK inhibitor 1 (Compound 27) is a BTK inhibitor (IC50 0.11 nM) with an inhibitory effect on B-cell activation in hWB with an IC50 of 2 nM. purity: 99%
Catalog No. 50-258-0224 Supplier TARGETMOL CHEMICALS INC Supplier No. T353305MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BTK inhibitor 1 (Compound 27) is a BTK inhibitor (IC50 0.11 nM) with an inhibitory effect on B-cell activation in hWB with an IC50 of 2 nM. purity: 99%