Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Btk inhibitor 2 (BGB-3111 analog) | 1558036-85-3 | MFCD31382019 | 99.6% | 431.49 g/mol | C24H25N5O3 | 25 MG
SDP

Catalog No. 5000245752
Encompass_Preferred

Btk inhibitor 2 (BGB-3111 analog) is a small-molecule inhibitor of Bruton's tyrosine kinase (BTK) described in patent US 20170224688 A1. It is supplied as a research reagent with reported high purity and is intended for in vitro biochemical and cellular studies.

  • Bruton's tyrosine kinase inhibitor for research use only.
  • Extracted from patent US 20170224688 A1.
  • Cas number 1558036-85-3.
  • Molecular formula C24H25N5O3.
  • Molecular weight 431.49 g/mol.
  • Reported purity 99.6%.
  • Available in small research pack sizes, including 25 mg.

Catalog No. 50-002-45752 Supplier Medchemexpress LLC Supplier No. HY10176625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Btk inhibitor 2 (BGB-3111 analog) is a small-molecule inhibitor of Bruton's tyrosine kinase (BTK) described in patent US 20170224688 A1. It is supplied as a research reagent with reported high purity and is intended for in vitro biochemical and cellular studies.

  • Bruton's tyrosine kinase inhibitor for research use only.
  • Extracted from patent US 20170224688 A1.
  • Cas number 1558036-85-3.
  • Molecular formula C24H25N5O3.
  • Molecular weight 431.49 g/mol.
  • Reported purity 99.6%.
  • Available in small research pack sizes, including 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.