Learn More
Grainger BTL COLIFORM STERILE PS 120 ML
Catalog No. NC9984800
Encompass_Preferred
Polystyrene Coliform Bottle, Sterile,120mL,38-400mm Cap Size,100/case,Mark indicating 100mL line makes sampling easier,Made of high-quality PS resin-won't fluoresce under black light
Catalog No. NC9984800 Supplier Grainger Supplier No. 3TRV5
Polystyrene Coliform Bottle, Sterile,120mL,38-400mm Cap Size,100/case,Mark indicating 100mL line makes sampling easier,Made of high-quality PS resin-won't fluoresce under black light