Learn More
Med Vet International Dispensing Bottle, 3mL, J1209, 12/Bag
Catalog No. 5000495734
Encompass_Preferred
These small volume dropper bottles are perfect for smaller pets or medications such as opthalmics. Also available in sterile packs• Bottle and cap made of slightly opaque, low density polyethylene• White cap made of polystyrene • Small drop volume inner nozzle3ml (0.1oz) bottle and cap 12/bag
Catalog No. 50-004-95734 Supplier Med Vet International Supplier No. JOR1209
These small volume dropper bottles are perfect for smaller pets or medications such as opthalmics. Also available in sterile packs• Bottle and cap made of slightly opaque, low density polyethylene• White cap made of polystyrene • Small drop volume inner nozzle3ml (0.1oz) bottle and cap 12/bag