Learn More
Med Vet International Amber Glass Dropper Bottle, 2oz/ 60mL, J1208B, 12/Box
Catalog No. 5000495657
Encompass_Preferred
Some medications interact with plastic bottles and plastic droppers. So a total glass product is preferred. 2.0oz (60ml) 12/box
Catalog No. 50-004-95657 Supplier Med Vet International Supplier No. JOR1208B
Some medications interact with plastic bottles and plastic droppers. So a total glass product is preferred. 2.0oz (60ml) 12/box