Learn More
Med Vet International Amber Glass Dropper Bottle, J1208, 0.5oz, 12 Count
Catalog No. 5000495785
Encompass_Preferred
Some medications interact with plastic bottles and plastic droppers. So a total glass product is preferred. 0.5oz (15ml) 12/box
Catalog No. 50-004-95785 Supplier Med Vet International Supplier No. JOR1208
Some medications interact with plastic bottles and plastic droppers. So a total glass product is preferred. 0.5oz (15ml) 12/box