Learn More
Grainger BTL FLIP-TOP DSPNSNG 240 ML HD
Catalog No. NC9205974
Encompass_Preferred
Flip-Top Dispensing Bottle, Refillable and Reusable so They are Ideal for Dispensing a Variety of Solutions - Even Harsh Chemical Solutions, Made of high-Density Polyethylene and Have Polypropylene Closures, 240mL
Catalog No. NC9205974 Supplier Grainger Supplier No. 3VEW9
Flip-Top Dispensing Bottle, Refillable and Reusable so They are Ideal for Dispensing a Variety of Solutions - Even Harsh Chemical Solutions, Made of high-Density Polyethylene and Have Polypropylene Closures, 240mL