Learn More
CHEMSCIENCE Square Glass Media/Storage Bottles, CLEAR, 1000 mL, BLUE CAPS,PK/10
Catalog No. NC2681229
Encompass_Preferred
Square bottles are easier to handle, take up less room on the shelf or in the autoclave (13% to 20%), and are perfect for mixing, sampling, and storing. White enamel graduations, a big marking point, and a glass ridge sculpted into the bottle to signify full capacity are among the features. A linerless, one-piece, autoclavable blue polypropylene plug seal cap with a drip-free pouring ring is included. The CS-CP45 series catalog contains a broad variety of alternative caps.
Catalog No. NC2681229 Supplier CHEMSCIENCE Supplier No. 274.602.04
Square bottles are easier to handle, take up less room on the shelf or in the autoclave (13% to 20%), and are perfect for mixing, sampling, and storing. White enamel graduations, a big marking point, and a glass ridge sculpted into the bottle to signify full capacity are among the features. A linerless, one-piece, autoclavable blue polypropylene plug seal cap with a drip-free pouring ring is included. The CS-CP45 series catalog contains a broad variety of alternative caps.