Learn More
Andwin Scientific BTL HDPE TRGR SPRY 16OZ 6/CS
SDP
Catalog No. NC9204487
Encompass_Preferred
The Adjustable Spray Bottles feature trigger-motion spray that dispenses a spray or stream of liquid from an adjustable nozzle. A strainer is attached to help keep solid particles from clogging the sprayer mechanism. The bottle has a volume capacity of 16 ounces. This is sold in a case of 6.
Catalog No. NC9204487 Supplier Andwin Scientific Supplier No. 7331X
The Adjustable Spray Bottles feature trigger-motion spray that dispenses a spray or stream of liquid from an adjustable nozzle. A strainer is attached to help keep solid particles from clogging the sprayer mechanism. The bottle has a volume capacity of 16 ounces. This is sold in a case of 6.