Learn More
HCL Labels 16 oz Pre-labeled GHS Spray Bottle Ethanol, Bottle Supplied Empty, Pack of 5
SDP
Catalog No. 501633584
Encompass_Preferred
Bottles, HCL Labels, Inc., 16oz, GHS Ethanol Spray Bottle, These bottles are pre-labeled with a GHS hazard communication chemical label. They are fully GHS compliant and include the necessary pictograms and signal words. Bottles are chemical resistant, supplied empty and come in packs of 5.
Catalog No. 50-163-3584 Supplier HCL Labels Supplier No. GHSBOT00045
Bottles, HCL Labels, Inc., 16oz, GHS Ethanol Spray Bottle, These bottles are pre-labeled with a GHS hazard communication chemical label. They are fully GHS compliant and include the necessary pictograms and signal words. Bottles are chemical resistant, supplied empty and come in packs of 5.