Learn More
Accuflow Systems BTL SAFETY CARRIER,BLACK EACH
Supplier:  Accuflow Systems B102
Encompass_Preferred
Resists most aqueous solutions, acids, bases - Handle molded as part of carrier - Carries liquids and ice without sweating Resists bacteria and fungi growth - Precision-molded to assure uniform appearance and quality - Centering ring in base holds bottle to assure clearance from wall. Height w/Bail: 16" Bucket Height: 6.5" Diameter 7.5" LARGE, BLACK
Catalog No. 50-109-4650