Learn More
Cole-Parmer Argos Technologies Disposable Bottle Top Aspirator System, SFCA Membrane, 0.45 um, 500mL, Sterile; 12/CS
Supplier:  Cole-Parmer 2440671
Encompass
Disposable bottletop aspirators provide a complete vacuum filtration solution for your separation and purification needs. All units come presterilized by gamma irradiation and certified nonpyrogenic, and are individually wrapped in easy-to-open packages. The large openings on the reservoir and funnel make for easy access. The funnel is made of clear polystyrene for easy viewing, and the bottle and filter adapter are made of 100% virgin polypropylene.
Catalog No. 50-196-1427