Learn More
Med Vet International Safety Labeled Wash Bottles, "Distilled Water" Labeled Wash Bottle, 1000mL (32 oz), J0817K, Each
Catalog No. 5000495150
Encompass_Preferred
Ensure leak proof dispensing of your common lab solutions. Polypropylene closure is 53mm W with a leak proof design. Easy-to-read and color-coded for content recognition and informative safety labeling:Printed bottle: Dept of Transportation (DOT)National fire protection (NFPA) codes and symbolsChemical abstract service number (CAS)Protective equipment symbols and target organ information All this information makes your workplace safer.
Catalog No. 50-004-95150 Supplier Med Vet International Supplier No. JOR817K
Ensure leak proof dispensing of your common lab solutions. Polypropylene closure is 53mm W with a leak proof design. Easy-to-read and color-coded for content recognition and informative safety labeling:Printed bottle: Dept of Transportation (DOT)National fire protection (NFPA) codes and symbolsChemical abstract service number (CAS)Protective equipment symbols and target organ information All this information makes your workplace safer.