Learn More
Chemglass Life Sciences Chemglass 500 mL LDPE Self-Venting Wash Bottle, Ethanol Labeled (Pack of 6)
SDP
Catalog No. NC2959218
Encompass_Preferred
This 500 mL low-density polyethylene (LDPE) wide-mouth wash bottle is pre-labeled for ethanol and is intended for safe short-term dispensing. It features a non-metallic self-venting closure, clear safety labeling, and is supplied in packs of six for laboratory use.
  • 500 mL capacity suitable for common laboratory dispensing tasks.
  • LDPE bottle construction provides break resistance and chemical compatibility with common solvents.
  • Wide mouth design makes filling and cleaning easier.
  • Non-metallic self-venting closure reduces solvent leakage risk while allowing pressure equalization.
  • Safety label includes CAS number, NFPA codes and symbols, protective equipment guidance, and hazard information.
  • Supplied in packs of six for convenient lab stocking.
Catalog No. NC2959218 Supplier Chemglass Life Sciences Supplier No. CG1131V004
This 500 mL low-density polyethylene (LDPE) wide-mouth wash bottle is pre-labeled for ethanol and is intended for safe short-term dispensing. It features a non-metallic self-venting closure, clear safety labeling, and is supplied in packs of six for laboratory use.
  • 500 mL capacity suitable for common laboratory dispensing tasks.
  • LDPE bottle construction provides break resistance and chemical compatibility with common solvents.
  • Wide mouth design makes filling and cleaning easier.
  • Non-metallic self-venting closure reduces solvent leakage risk while allowing pressure equalization.
  • Safety label includes CAS number, NFPA codes and symbols, protective equipment guidance, and hazard information.
  • Supplied in packs of six for convenient lab stocking.